missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569457
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 100%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 85%; Rat: 93%; Yeast: 100%.
Natural killer cells express multiple calcium-dependent lectin-like receptors, such as CD94 and NKG2D, that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC2 is a C-type lectin-like receptor expressed in myeloid cells and NK cells.
Specifications
| CLEC1B | |
| Polyclonal | |
| Unconjugated | |
| CLEC1B | |
| 1810061I13Rik; Clec1b; CLEC2; CLEC-2; CLEC2B; CLEC9A; c-type lectin 1 member B; C-type lectin domain containing 9A; C-type lectin domain family 1 member B; C-type lectin domain family 1, member b; C-type lectin domain family 9 member A; C-type lectin domain family 1 member B; C-type lectin-like receptor 2; C-type lectin-like receptor-2; PRO1384; QDED721; RGD1563517; UNQ721/PRO1384 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51266, 56760 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9JL99, Q9P126 | |
| CLEC1B | |
| synthetic peptide directed towards the middle region of human CLEC1B (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction