missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLEC2B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5116747

Product Code. 17289243

  • € 506.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Samples: Rat spleen Immunogen sequence: IGFQNKCYYF SKEEGDWNSS KYNCSTQHAD LTIIDNIEEM NFLRRYKCSS DHWIGLKMAK NRTGQWVDGA TFTKSFGMRG S.

CLEC2B encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell activation antigen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CLEC2B
Polyclonal
Unconjugated
CLEC2B
Activation-induced C-type lectin; AICL; CLEC2B; CLECSF2; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 2 (activation-induced); C-type lectin domain family 2 member B; C-type lectin domain family 2, member B; C-type lectin superfamily member 2; HP10085; IFN-alpha2b-inducing related protein 1; IFN-alpha-2b-inducing-related protein 1; IFNRG1
Recombinant fusion protein containing a sequence corresponding to amino acids 40-120 of human CLEC2B (NP_0051182).
100 μL
Primary
Rat
Antibody
IgG
ELISA, Western Blot
1.56 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q92478
Rabbit
Affinity Chromatography
RUO
9976
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.