missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC2L Antibody (2326G), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10204K
This item is not returnable.
View return policy
Description
CLEC2L Monoclonal antibody specifically detects CLEC2L in Human samples. It is validated for Serum
Specifications
| CLEC2L | |
| Monoclonal | |
| DyLight 488 | |
| C-type lectin domain family 2 member L, C-type lectin domain family 2, member L | |
| Human embryonic kidney cell, HEK293-derived human CLEC2L protein, Ser90-Thr214, Accession # P0C7M8-1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2326G | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 154790 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction