missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102996
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CLEC3B.
Tetranectin (TN) is a serum and tissue protein, a C-type lectin, which binds to Ca++. It is a homotrimer of monomers each with a mass of 20 kDa, plasma or serum concentrations of TN are found to be approximately 10 mg/l. In vitro, TN can bind to kringle 4 of plasminogen and enhance the activation of plaminogen to plasmin, catalyzed by tissue plasminogen activator in the presence of poly-D-lysine. TN is best known as a prognostic marker in ovarian cancer.
Specifications
| CLEC3B | |
| Polyclonal | |
| Unconjugated | |
| Clec3b | |
| CLEC3B; C-type lectin domain family 3 member B; C-type lectin domain family 3, member b; LOC100912012; Plasminogen kringle 4-binding protein; tetranectin; tetranectin (plasminogen binding protein); tetranectin (plasminogen-binding protein); tetranectin-like; TN; TNA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100912012, 21922, 7123 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05452, P43025 | |
| Clec3b | |
| A synthesized peptide derived from human CLEC3B(Accession P05452), corresponding to amino acid residues M79-A129. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction