missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LDH-Cytotoxicity Assay Kit (Colorimetric)

Product Code. 18616366 Shop All Bio Techne Products
Click to view available options
Quantity:
400 Assays
Unit Size:
400 tests
This item is not returnable. View return policy

Product Code. 18616366

Brand: Novus Biologicals™ NBP254847

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Assay Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
Quantity 400 Assays
Species Human, Mouse, Rat, Mammal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.