missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC4D/CLECSF8 Antibody (9B9), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18866AF350
This item is not returnable.
View return policy
Description
CLEC4D/CLECSF8 Monoclonal antibody specifically detects CLEC4D/CLECSF8 in Human, Primate samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| CLEC4D/CLECSF8 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| CLEC6, CLEC-6, CLECSF8Mpcl, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 8, C-type lectin domain family 4 member D, C-type lectin domain family 4, member D, C-type lectin receptor, C-type lectin superfamily member 8, C-type lectin-like receptor 6, macrophage C-type lectin, MCL, MGC40078, MPCL | |
| CLEC4D/CLECSF8 ectodomain fused with human Fc | |
| 0.1 mL | |
| Primary | |
| Human, Primate | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation | |
| 9B9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 338339 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction