missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC4D Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547403
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) CLECSF9 and rhCLEC4F is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
Specifications
| CLEC4D | |
| Polyclonal | |
| Unconjugated | |
| CLEC4D | |
| CD368; Clec4d; CLEC6; CLEC-6; CLECSF8; CLECSF8Mpcl; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 8; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 8; C-type lectin domain family 4 member D; C-type lectin domain family 4, member d; C-type lectin receptor; C-type lectin superfamily member 8; C-type lectin, superfamily member 8; C-type lectin-like receptor 6; macrophage C-type lectin; macrophage-restricted C-type lectin; Mcl; MGC40078; Mpcl; superfamilymember 8 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 338339 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8WXI8 | |
| CLEC4D | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC4D/CLECSF8 Gly52-Asn215. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction