missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC4D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116974
This item is not returnable.
View return policy
Description
Positive Samples: Mouse spleen, Rat lung Immunogen sequence: CLVTHHNFSR CKRGTGVHKL EHHAKLKCIK EKSELKSAEG STWNCCPIDW RAFQSNCYFP LTDNKTWAES ERNCSGMGAH LMTISTEAEQ NFIIQFLDRR LSYFLGLRDE NAKGQWRWVD QTPFNPRRVF WHKNEPDNSQ GENCVVLVYN QDKWAWNDVP CNFEASRICK IPGTTLN.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
Specifications
| CLEC4D | |
| Polyclonal | |
| Unconjugated | |
| CLEC4D | |
| CD368; Clec4d; CLEC6; CLEC-6; CLECSF8; CLECSF8Mpcl; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 8; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 8; C-type lectin domain family 4 member D; C-type lectin domain family 4, member d; C-type lectin receptor; C-type lectin superfamily member 8; C-type lectin, superfamily member 8; C-type lectin-like receptor 6; macrophage C-type lectin; macrophage-restricted C-type lectin; Mcl; MGC40078; Mpcl; superfamilymember 8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17474, 338339, 362432 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.43 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q69FH1, Q8WXI8, Q9Z2H6 | |
| CLEC4D | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 39-215 of human CLEC4D (NP_5251262). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction