missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC4E Antibody (2455C), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8995MFV500
This item is not returnable.
View return policy
Description
CLEC4E Monoclonal antibody specifically detects CLEC4E in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CLEC4E | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Immunology | |
| 26253 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 2455C | |
| Flow Cytometry, CyTOF-ready | |
| CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9 | |
| Chinese Hamster Ovary cell line, CHO-derived recombinant human CLEC4E, Arg41-Leu219, Accession # Q9ULY5 | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction