missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC4E Antibody (OTI2A8), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71793AF488
This item is not returnable.
View return policy
Description
CLEC4E Monoclonal specifically detects CLEC4E in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| CLEC4E | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CLEC4E | |
| Full length recombinant protein of human CLEC4E (NP_055173) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2A8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, mincle, MINCLEC-type lectin superfamily member 9 | |
| Mouse | |
| Affinity Purified | |
| Immunology | |
| 26253.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction