missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLEC5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111388
This item is not returnable.
View return policy
Description
Immunogen sequence: PSPNGFITTR SYGTVCPKDW EFYQARCFFL STSESSWNES RDFCKGKGST LAIVNTPEKL KFLQD Highest antigen sequence indentity to the following orthologs: Rat - 66%, Mouse - 62%.
Functions as a positive regulator of osteoclastogenesis. Cell surface receptor that signals via TYROBP. Regulates inflammatory responses. Acts as a key regulator of synovial injury and bone erosion during autoimmune joint inflammation. Critical macrophage receptor for dengue virus serotypes 1-4. The binding of dengue virus to CLEC5A triggers signaling through the phosphylation of TYROBP, this interaction does not result in viral entry, but stimulates proinflammatory cytokine release.
Specifications
| CLEC5A | |
| Polyclonal | |
| Unconjugated | |
| CLEC5A | |
| CLEC5A; Clecsf5; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 5; C-type lectin domain family 5 member A; C-type lectin domain family 5, member a; C-type lectin superfamily member 5; Ly100; Mdl1; MDL-1; myeloid DAP12-associating lectin 1; myeloid DAP12-associating lectin-1; type II transmembrane protein MDL-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23601 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NY25 | |
| CLEC5A | |
| Recombinant Protein corresonding to Human CLEC5A. Recombinant protein control fragment (Product #RP-108400). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction