missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC9a Antibody (1F6), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-08840AF350
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal antibody specifically detects CLEC9a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| CLEC9a | |
| Monoclonal | |
| Alexa Fluor 350 | |
| C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| RBL-2H3 cells expressing Mouse CLEC9a fused to an HA epitope. | |
| 100 μg | |
| Immunology, Stem Cells | |
| 283420 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| 1F6 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction