missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC9a Antibody (30L2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21999AF488
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal specifically detects CLEC9a in Human samples. It is validated for Flow Cytometry.
Specifications
| CLEC9a | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Q6UXN8 | |
| CLEC9A | |
| Human recombinant CLEC9a | |
| 0.1 mL | |
| Immunology, Stem Cells | |
| 283420 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 30L2 | |
| Flow Cytometry | |
| C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction