missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC9a Antibody (30L2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21999H
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal antibody specifically detects CLEC9a in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CLEC9a | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 30L2 | |
| Flow Cytometry, CyTOF-ready | |
| C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| Human recombinant CLEC9a | |
| 0.1 mL | |
| Immunology, Stem Cells | |
| 283420 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction