missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC9a Antibody (397) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50168-0.025ml
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal antibody specifically detects CLEC9a in Mouse samples. It is validated for Western Blot
Specifications
| CLEC9a | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000 | |
| C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| RBL-2H3 cells expressing mouse CLEC9a fused to an HA epitope | |
| 0.025 mL | |
| Immunology, Stem Cells | |
| 283420 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot | |
| 397 | |
| Unconjugated | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction