missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC9a Antibody (7H11), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB67761C
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal antibody specifically detects CLEC9a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| CLEC9a | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 7H11 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| RBL-2H3 rat basophilic leukemia cell line expressing mouse CLEC9a fused to an HA epitope, Accession # Q8BRU4 | |
| 0.1 mL | |
| Immunology, Stem Cells | |
| 283420 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction