missing translation for 'onlineSavingsMsg'
Learn More

Clemastine Fumarate, TRC

Product Code. 30359787
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359787 25 mg -
30349437 100 mg -
30354326 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359787 Supplier Toronto Research Chemicals Supplier No. TRCC56850025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14976-57-9
Molecular Formula C21 H26 Cl N O . C4 H4 O4
Molecular Weight (g/mol) 459.96
Synonym Clemastine fumarate, (2R)-2-[2-[(R)-1-(4-Chlorophenyl)-1-phenylethoxy]ethyl]-1-methylpyrrolidine (E)-butenedioate, Pyrrolidine, 2-[2-[(1R)-1-(4-chlorophenyl)-1-phenylethoxy]ethyl]-1-methyl-, (2R)-, (2E)-2-butenedioate (1:1), Clemastine Fumarate, Pyrrolidine, 2-[2-[(p-chloro-α-methyl-α-phenylbenzyl)oxy]ethyl]-1-methyl-, (+)-, fumarate (1:1) (8CI), Pyrrolidine, 2-[2-[1-(4-chlorophenyl)-1-phenylethoxy]ethyl]-1-methyl-, [R-(R*,R*)]-, (E)-2-butenedioate (1:1), (+)-2-[2-[(p-Chloro-α-methyl-α-phenylbenzyl)oxy]ethyl]-1-methylpyrrolidine fumarate, Agasten, Aloginan, Alphamin, Anhistan, Bravegyl, Clemastine fumarate, Clemastine hydrogen fumarate, Fuluminol, HS 592, Inbestan, Kinotomin, Lacretin, Lecasol, Maikohis, Mallermin F, Marsthine, Masletine, Meclastine fumarate, Mecloprodine, Piloral, Reconin, Tavegil, Tavegyl, Tavist, Telgin, Telgin G, Trabest, Xolamin
IUPAC Name (E)-but-2-enedioic acid;(2R)-2-[2-[(1R)-1-(4-chlorophenyl)-1-phenylethoxy]ethyl]-1-methylpyrrolidine
SMILES CN1CCC[C@@H]1CCO[C@](C)(c2ccccc2)c3ccc(Cl)cc3.OC(=O)\C=C\C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H26ClNO.C4H4O4/c1-21(17-7-4-3-5-8-17,18-10-12-19(22)13-11-18)24-16-14-20-9-6-15-23(20)2;5-3(6)1-2-4(7)8/h3-5,7-8,10-13,20H,6,9,14-16H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/t20-,21-;/m1./s1
Formula Weight 459.1813
Chemical Name or Material Clemastine Fumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.