missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clervonafusp Alfa Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554860
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Clervonafusp alfa is a fusion protein that targets and binds to the transferrin receptor 1 (TfR1) on the blood-brain barrier, facilitating the delivery of the enzyme iduronate-2-sulfatase (I2S) into the central nervous system.
Specifications
| Clervonafusp Alfa Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HNP36GAA | |
| Recombinant Human ENT2/SLC29A2 and GAA/Aglucosidase alfa. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction