Learn More
Clethodim, TRC
Specifications
Specifications
| Chemical Name or Material | Clethodim |
| CAS | 99129-21-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H26 Cl N O3 S |
| Quantity | 50 mg |
| Synonym | 2-Cyclohexen-1-one, 2-[1-[[[(2E)-3-chloro-2-propenyl]oxy]imino]propyl]-5-[2-(ethylthio)propyl]-3-hydroxy- (9CI), 2-[1-[[[(2E)-3-Chloro-2-propen-1-yl]oxy]imino]propyl]-5-[2-(ethylthio)propyl]-3-hydroxy-2-cyclohexen-1-one, Centurion, Centurion 240, Centurion Plus, Clethodim, E-Clethodim, Ogive, Prism, Prism (pesticide), RE 45601, Select, Select MAX, Select Super 120EC, Trigger, Volunteer |
| InChI Formula | InChI=1S/C17H26ClNO3S/c1-4-14(19-22-8-6-7-18)17-15(20)10-13(11-16(17)21)9-12(3)23-5-2/h6-7,12-13,20H,4-5,8-11H2,1-3H3/b7-6+,19-14+ |
| SMILES | CCSC(C)CC1CC(=C(\C(=N\OC\C=C\Cl)\CC)C(=O)C1)O |
| IUPAC Name | 2-[(E)-N-[(E)-3-chloroprop-2-enoxy]-C-ethylcarbonimidoyl]-5-(2-ethylsulfanylpropyl)-3-hydroxycyclohex-2-en-1-one |
| Molecular Weight (g/mol) | 359.91 |
| Show More |