missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ clfA Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144498
This item is not returnable.
View return policy
Description
clfA Polyclonal Antibody for Western Blot, ELISA
Cell surface-associated protein implicated in virulence. Promotes bacterial attachment exclusively to the gamma-chain of human fibrinogen. Induces formation of bacterial clumps (By similarity).
Specifications
| clfA | |
| Polyclonal | |
| Unconjugated | |
| clfA; Clumping factor A | |
| Recombinant Staphylococcus aureus Clumping factor A (clfA), partial (228-558AA). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.9 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction