missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLGN Antibody (9C8G10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37395
This item is not returnable.
View return policy
Description
CLGN Monoclonal specifically detects CLGN in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CLGN | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| calmegin | |
| Mouse | |
| 70 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 9C8G10 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CLGN | |
| Purified recombinant fragment of human CLGN (AA: 249-405) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 1047 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction