missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLIC1 Polyclonal Antibody

Product Code. 15564552
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15564552 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15564552 Supplier Invitrogen™ Supplier No. PA541004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: GQLPFLLYGT EVHTDTNKIE EFLEAVLCPP RYPKLAALNP ESNTAGLDIF Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 1 is a member of the p64 family; the protein localizes principally to the cell nucleus and exhibits both nuclear and plasma membrane chloride ion channel activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIC1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CLIC1
Gene Accession No. O00299, Q9Z1Q5
Gene Alias Chloride channel ABP; chloride intracellular channel 1; chloride intracellular channel protein; chloride intracellular channel protein 1; chloride intracellular channel protein 1 {ECO:0000250; Clcp; CLIC1; clic1 {ECO:0000312; CLIC1 protein; CLIC1; NCC27; DADB-110M10.1; EMBL:CAE83985.1, ECO:0000312; EMBL:CAE83985.1}; FLJ94671; G6; hRNCC; hypothetical protein LOC324481; NCC27; Nuclear chloride ion channel 27; nuclear chloride ion channel protein; p64CLCP; regulatory nuclear chloride ion channel protein; RGD:1303043}; RNCC protein; UniProtKB:O00299, ECO:0000312; wu:fc30e04; zgc:77044
Gene Symbols CLIC1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CLIC1 (aa 62-111).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114584, 1192
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.