missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CLIC4 Monoclonal antibody specifically detects CLIC4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | CLIC4 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:500, ELISA Recommended starting concentration is 1 μg/mL |
| Formulation | PBS (pH 7.3), 50% glycerol, 0.05% BSA |
| Gene Alias | chloride intracellular channel 4, chloride intracellular channel 4 like, chloride intracellular channel protein 4, CLIC4L, DKFZp566G223, FLJ38640, H1, huH1, Intracellular chloride ion channel protein p64H1, MTCLIC, p64H1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human CLIC4 (NP_039234.1).,, Sequence:, MALSMPLNGLKEEDKEPLIELFVKAGSDGESIGNCPFSQRLFMILWLKGVVFSVTTVDLKRKPADLQNLAPGTHPPFITFNSEVKTDVNKIEEFLEEVLC |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?