Learn More
Nickel Electro™ Clifton™ Stainless Steel Tank
Stainless Steel Tank providing an external vessel of circulating temperature controlled water.
€ 315.00 - € 457.00
Specifications
Type | Stainless Steel Tank |
---|---|
For Use With (Application) | Laboratory Use |
Material | Stainless Steel, Antibacterial Paint Finish |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11756587
|
Nickel Electro™
NE4-8U |
8 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11726587
|
Nickel Electro™
NE4-14U |
14 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11736587
|
Nickel Electro™
NE4-22U |
22 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Double-walled construction
- External surfaces have an anti-bacterial paint finish
- Inner tank is a bright stainless steel.
- While in use, submerge Inlet and outlet hoses under water from NE6-FH Clifton Flow Heater.
Specifications
Stainless Steel Tank | |
Laboratory Use | |
Stainless Steel, Antibacterial Paint Finish |