missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLINT1 Monoclonal Antibody (OTI9E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805981S
This item is not returnable.
View return policy
Description
CLINT1 Monoclonal antibody specifically detects CLINT1 in Human samples. It is validated for Western BlotSpecifications
| CLINT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14677 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9685 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI9E1 | |
| Unconjugated | |
| CLINT1 | |
| AI642036; AW537616; C530049I24Rik; clathrin interacting protein localized in the trans-Golgi region; clathrin interactor 1; Clathrin interactor 1-like protein; Clathrin-interacting protein localized in the trans-Golgi region; Clint; Clint1; ENTH; ENTH domain containing protein; enthoprotin; Epn4; EPNR; epsin 4; epsin-4; epsinR; Epsin-related protein; F630206G17Rik; KIAA0171; mKIAA0171 | |
| Human recombinant protein fragment corresponding to amino acids 1-290 of human CLINT1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction