Learn More
CLINT1 Mouse anti-Human, Clone: OTI8C1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF805980
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Binds to membranes enriched in phosphatidylinositol 4,5- bisphosphate (PtdIns(4,5)P2). May have a role in transport via clathrin-coated vesicles from the trans-Golgi network to endosomes. Stimulates clathrin assembly.Specifications
| CLINT1 | |
| Monoclonal | |
| Unconjugated | |
| CLINT1 | |
| CLINT, ENTH, EPN4, EPNR | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9685 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI8C1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q14677 | |
| CLINT1 | |
| Human recombit protein fragment corresponding to amino acids 1-290 of human CLINT1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.