Learn More
Invitrogen™ CLN2 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA547275
Product Code. 15597762
  • € 530.00 / 100µg
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the sedolisin family of serine proteases. The protease functions in the lysosome to cleave N-terminal tripeptides from substrates, and has weaker endopeptidase activity. It is synthesized as a catalytically-inactive enzyme which is activated and auto-proteolyzed upon acidification. Mutations in this gene result in late-infantile neuronal ceroid lipofuscinosis, which is associated with the failure to degrade specific neuropeptides and a subunit of ATP synthase in the lysosome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CLN2
Polyclonal
Unconjugated
TPP1
cell growth-inhibiting gene 1 protein; ceroid-lipofuscinosis, neuronal 2; CLN 2; CLN2; GIG 1; GIG1; growth-inhibiting protein 1; LPIC; lysosomal pepstatin insensitive protease; lysosomal pepstatin-insensitive protease; MGC21297; SCAR7; TPP 1; TPP I; Tpp1; TPP-1; TPPI; TPP-I; Tripeptidyl aminopeptidase; tripeptidyl peptidase 1; tripeptidyl peptidase I; tripeptidyl peptidase-I; tripeptidyl-peptidase 1; Tripeptidyl-peptidase I; UNQ267/PRO304
Goat
Antigen affinity chromatography
RUO
1200
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
O14773
TPP1
Mouse myeloma cell line NS0-derived recombinant human Tripeptidyl-PeptidaseI/TPP1 Ser20-Pro563.