missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLN8 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99467FR
This item is not returnable.
View return policy
Description
CLN8 Polyclonal antibody specifically detects CLN8 in Human samples. It is validated for Western Blot
Specifications
| CLN8 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| C8orf61, ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation), EPMR | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CLN8. | |
| 0.1 mL | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 2055 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction