missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLN8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99467-50ul
This item is not returnable.
View return policy
Description
CLN8 Polyclonal antibody specifically detects CLN8 in Human samples. It is validated for Western Blot
Specifications
| CLN8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C8orf61, ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation), EPMR | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CLN8. | |
| 50 μg | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 2055 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?