missing translation for 'onlineSavingsMsg'
Learn More

Clobetasol 17-Propionate-d5, TRC

Product Code. 30358019
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358019 10 mg -
30349325 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30358019 Supplier Toronto Research Chemicals Supplier No. TRCC58350210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C25H27D5ClFO5
Molecular Weight (g/mol) 472
Synonym (11β,16β)-21-Chloro-9-fluoro-11-hydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione-d5, 21-Chloro-21-deoxybetamethasone 17-Propionate-d5, Clobederm-d5, Clobesol-d5, Clobex-d5, Dermoval-d5, Dermovate-d5, Dermoxin-d5, Dermoxinale-d5, GR 2/925-d5, Karison-d5, Olux-d5, Skin Cap-d5, Temovate-d5
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-17-(2-chloroacetyl)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] 2,2,3,3,3-pentadeuteriopropanoate
SMILES O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2(F)[C@@H](O)C[C@@]4(C)[C@@]3([H])C[C@H](C)[C@]4(OC(C([2H])([2H])C([2H])([2H])[2H])=O)C(CCl)=O)=C1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C25H32ClFO5/c1-5-21(31)32-25(20(30)13-26)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,27)19(29)12-23(18,25)4/h8-9,11,14,17-19,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1/i1D3,5D2
Percent Purity >95%
Chemical Name or Material Clobetasol 17-Propionate-d5
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.