missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Antibody (8F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51610
Additional Details : Weight : 0.00970kg
Description
CLOCK Monoclonal specifically detects CLOCK in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CLOCK | |
Monoclonal | |
Unconjugated | |
BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein | |
Mouse | |
95 kDa | |
0.1 mL | |
Primary | |
Human | |
Ascites |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
8F7 | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
CLOCK | |
Purified recombinant fragment of human CLOCK expressed in E. Coli. | |
Unpurified | |
RUO | |
9575 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
For Research Use Only