missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Antibody (OTI2H7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46311
Additional Details : Weight : 0.00970kg
Descrizione
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
CLOCK Monoclonal specifically detects CLOCK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifica
CLOCK | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein | |
Mouse | |
95.1 kDa | |
0.1 mL | |
Cancer, Chromatin Research, Circadian Rhythm, Hypoxia, Neuroendocrinology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision | |
9575 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Immunohistochemistry (Paraffin), Immunohistochemistry | |
OTI2H7 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
CLOCK | |
Human recombinant protein fragment corresponding to amino acids 1-285 of human CLOCK (NP_004889) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
For Research Use Only