missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Monoclonal Antibody (OTI4A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804825S
This item is not returnable.
View return policy
Description
CLOCK Monoclonal antibody specifically detects CLOCK in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CLOCK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P63316 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7134 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4A6 | |
| Unconjugated | |
| Tnnc1 | |
| 5330400M04Rik; bHLHe8; circadian locomoter output cycles kaput; circadian locomoter output cycles kaput protein; circadian locomoter output cycles protein kaput; circadian locomotor output cycles kaput; class E basic helix-loop-helix protein 8; CLOCK; clock circadian regulator; clock homolog; hCLOCK; I79_020252; KAT13D; KIAA0334; mCLOCK; rCLOCK | |
| Full length human recombinant protein of human TNNC1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction