missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Monoclonal Antibody (OTI4E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804781
This item is not returnable.
View return policy
Description
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1.Specifications
| CLOCK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15516 | |
| CLOCK | |
| Human recombit protein fragment corresponding to amino acids 1-285 of human CLOCK produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4E12 | |
| Unconjugated | |
| CLOCK | |
| KAT13D, bHLHe8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9575 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction