Learn More
CLOCK Mouse anti-Human, Clone: OTI4E12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804781
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1.Specifications
| CLOCK | |
| Monoclonal | |
| Unconjugated | |
| CLOCK | |
| KAT13D, bHLHe8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9575 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4E12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O15516 | |
| CLOCK | |
| Human recombit protein fragment corresponding to amino acids 1-285 of human CLOCK produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.