Learn More
STEMCELL Technologies ClonaCell™-CHO CD Liquid Medium
Product Code. 15335395
Encompass
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 15335395
Brand: STEMCELL Technologies 03817
Chemically defined, animal component-free, serum-free, protein-free, glutamine-free liquid medium for culture of CHO cells
  • ClonaCell™-CHO CD Liquid Medium is a general purpose liquid medium for culture of suspension-adapted CHO cells
  • This chemically defined liquid medium is protein-free and animal component-free
  • It does not contain L-glutamine, selection agents, antibiotics, or phenol red
  • ClonaCell™-CHO CD Liquid Medium is compatible with a variety of selection systems, including dihydrofolate reductase (DHFR) and glutamine synthetase (GS)
Specifications
Cell Line CHO
For Use With (Application) Cell Culture
Quantity 500 mL