Learn More
STEMCELL Technologies ClonaCell™-TCS/G418 Kit
Product Code. 17128211
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17128211 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17128211 Supplier STEMCELL Technologies Supplier No. 03822
Complete kit containing ClonaCell™-TCS medium and antibiotic G418
  • This kit is used for selection and cloning applications where the G418 antibiotic is used as the selective agent
  • ClonaCell™-TCS Medium is a serum-containing, methylcellulose-based semi-solid medium that can be used for selection and cloning of a variety of suspension-adapted and adherent cell lines, such as BHK-1, CHO-K1, CHO-S, CHO-DG44, and HEK-293
  • This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA)
  • The G418 antibiotic is suitable for use in cell culture for the selection and maintenance of G418-resistant clones when using ClonaCell™-TCS Medium
Specifications
Cell Line CHO Cells, HEK-293, Other
For Use With (Application) Cell Culture; Semi-Solid Cloning
Quantity 1 Kit