missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Closantel Sodium Salt, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Closantel Sodium |
| CAS | 61438-64-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H13 Cl2 I2 N2 O2 . Na |
| Quantity | 5 g |
| Synonym | Benzamide, N-[5-chloro-4-[(4-chlorophenyl)cyanomethyl]-2-methylphenyl]-2-hydroxy-3,5-diiodo-, sodium salt (1:1), Benzamide, N-[5-chloro-4-[(4-chlorophenyl)cyanomethyl]-2-methylphenyl]-2-hydroxy-3,5-diiodo-, monosodium salt (9CI) |
| InChI Formula | InChI=1S/C22H14Cl2I2N2O2.Na/c1-11-6-15(17(10-27)12-2-4-13(23)5-3-12)18(24)9-20(11)28-22(30)16-7-14(25)8-19(26)21(16)29;/h2-9,17,29H,1H3,(H,28,30);/q;+1/p-1 |
| SMILES | [Na+].Cc1cc(C(C#N)c2ccc(Cl)cc2)c(Cl)cc1NC(=O)c3cc(I)cc(I)c3[O-] |
| IUPAC Name | sodium;2-[[5-chloro-4-[(4-chlorophenyl)-cyanomethyl]-2-methylphenyl]carbamoyl]-4,6-diiodophenolate |
| Molecular Weight (g/mol) | 685.06 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction