missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clostridia Monoclonal Antibody (J44D)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518193
This item is not returnable.
View return policy
Description
This antibody is specific for glutamate dehydrogenase of Clostridia species. It is reactive with recombinant GDH and native GDH from C. difficile cultures. Purity is >90%.
Clostridia are a highly polyphyletic class of Firmicutes, including Clostridium and other similar genera. Clostridia are distinguished from the Bacilli by lacking aerobic respiration. Clostridia are obligate anaerobes and oxygen is toxic to them. Diseases caused by Clostridia include botulism from C. botulinum as a result of exposure to botulinum toxin.
Specifications
| Clostridia | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA | |
| J44D | |
| Unconjugated | |
| Clostridium | |
| Clostridia recombinant protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction