missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clostridium botulinum Toxin A VHH Recombinant Alpaca Monoclonal Antibody (SAA1450)
Alpaca Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554315
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Clostridium botulinum is a gram-positive, anaerobic motile bacteria normally found in soil. C. botulinum is a potential threat to both humans and animals through contaminated food because of the potent neurotoxin they produce. C. botulinum bacteria are categorized within 7 strains A-G based on the characteristics of their toxin. A,B, E and F strains are known to cause food poisoning in humans.
Specifications
| Clostridium botulinum Toxin A VHH | |
| Recombinant Monoclonal | |
| 1.05 mg/mL | |
| PBS with no preservative | |
| Alpaca | |
| Metal-chelate chromatography | |
| RUO | |
| Bacteria | |
| Antibody | |
| VHH |
| ELISA, Western Blot | |
| SAA1450 | |
| Unconjugated | |
| botulinum; botulinum toxin; C. botulinum | |
| Recombinant Clostridium botulinum botA/BOTOX. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction