Learn More
Invitrogen™ Clostridium difficile Toxin A Monoclonal Antibody (5147)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547549
Description
Specificity: Specific for toxin A of Clostridium difficile. Negative vs. toxin B. Best ELISA pairs (capture/conjugate): Clostridium difficile Toxin A Monoclonal antibody (Product # MA1-7416)/Clostridium difficile Toxin A Monoclonal antibody (Product # MA5-47549); Clostridium difficile Toxin A Polyclonal antibody (Product # MA1-7414)/Clostridium difficile Toxin A Monoclonal antibody (Product # MA5-47549). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Clostridium difficile is a species of bacteria of the genus Clostridium which are Gram-positive, anaerobic, spore-forming rods (bacillus). C.difficile is the most significant cause of pseudomembranous colitis, a severe infection of the colon.
Specifications
| Clostridium difficile Toxin A | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA | |
| 5147 | |
| Unconjugated | |
| TcdA | |
| Proprietary sequence. | |
| 100 μg | |
| Primary | |
| 4°C or -20°C if preferred | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.