missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clostridium Perfringens Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-62474AF532
This item is not returnable.
View return policy
Description
Clostridium Perfringens Polyclonal antibody specifically detects Clostridium Perfringens in Bacteria samples. It is validated for ELISA
Specifications
| Clostridium Perfringens | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Bacteria | |
| Purified |
| ELISA | |
| Alexa Fluor 532 | |
| C. perfringens | |
| Clostridium perfringens ATCC 13124 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction