missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clostridium Tetani Toxoid Antibody (6E7), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23554JF646
This item is not returnable.
View return policy
Description
Clostridium Tetani Toxoid Monoclonal antibody specifically detects Clostridium Tetani Toxoid in Bacteria samples. It is validated for ELISA
Specifications
| Clostridium Tetani Toxoid | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Bacteria | |
| Purified |
| ELISA | |
| 60000000 | |
| ELISA | |
| Clostridium tetani toxoid, Tentoxylysin, Tetanus toxin, tetX | |
| Tetanus toxoid adsorbed to aluminum hydroxide gel. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction