missing translation for 'onlineSavingsMsg'
Learn More

Cloxacillin sodium monohydrate, MedChemExpress

Product Code. 16462146
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16462146

Brand: Medchem Express HYB0466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cloxacillin sodium monohydrate is an orally active antibacterial agent and β-lactamase inhibitor with an IC50 of 0.04 μM. Cloxacillin sodium monohydrate can suppress the S. aureus-induced inflammatory response by inhibiting the activation of MAPKs, NF-κB and NLRP3-related proteins.

Cloxacillin sodium monohydrate is an orally active antibacterial agent and β-lactamase inhibitor with an IC50 of 0.04 μM. Cloxacillin sodium monohydrate can suppress the S. aureus-induced inflammatory response by inhibiting the activation of MAPKs, NF-κB and NLRP3-related proteins.

Specifications

Chemical Name or Material Cloxacillin sodium monohydrate
Physical Form Solid
CAS 7081-44-9
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C19H19ClN3NaO6S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (210.14 mM; Need ultrasonic) ∣H2O : 50 mg/mL (105.07 mM; Need ultrasonic)
SMILES O=C(C(C(C(C=CC=C1)=C1Cl)=NO2)=C2C)N[C@H]3[C@](SC(C)(C)[C@@H]4C(O[Na])=O)([H])N4C3=O.O
Molecular Weight (g/mol) 475.88
Formula Weight 475.88
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.