missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLP1/HEAB Antibody (SR1834), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22477-100ul
This item is not returnable.
View return policy
Description
CLP1/HEAB Monoclonal antibody specifically detects CLP1/HEAB in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| CLP1/HEAB | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10978 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| SR1834 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| CLP1, cleavage and polyadenylation factor I subunit, homolog (S. cerevisiae), EC 2.7.1.78, hClp1, HEAB, HEABATP/GTP-binding protein, homolog of yeast CFIA subunit Clp1p, Polynucleotide kinase Clp1, polyribonucleotide 5'-hydroxyl-kinase Clp1, Pre-mRNA cleavage complex II protein Clp1 | |
| A synthesized peptide derived from human CLP1/HEAB (Uniprot #: Q92989) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction