missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLP1 Monoclonal Antibody (OTI2D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809648S
This item is not returnable.
View return policy
Description
CLP1 Monoclonal antibody specifically detects CLP1 in Human samples. It is validated for Western BlotSpecifications
| CLP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14956 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10457 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2D2 | |
| Unconjugated | |
| GPNMB | |
| AI462438; ATP/GTP-binding protein; cleavage and polyadenylation factor I subunit 1; CLP1; CLP1, cleavage and polyadenylation factor I subunit; CLP1, cleavage and polyadenylation factor I subunit, homolog; HAMAP-Rule:MF_03035}; hClp1; Heab; homolog of yeast CFIA subunit Clp1p; polyadenylation factor Clp1; polyadenylation factor Clp1 {ECO:0000255; Polynucleotide kinase Clp1; polynucleotide kinase Clp1 {ECO:0000255; polyribonucleotide 5'-hydroxyl-kinase Clp1; polyribonucleotide 5'-hydroxyl-kinase Clp1 {ECO:0000255; Pre-mRNA cleavage complex II protein Clp1; pre-mRNA cleavage complex II protein Clp1 {ECO:0000255; RGD1307679 | |
| Human recombinant protein fragment corresponding to amino acids 508-572 of human GPNMB produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction