missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLP24/TMEM204 Antibody (652424) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6747Y
This item is not returnable.
View return policy
Description
CLP24/TMEM204 Monoclonal antibody specifically detects CLP24/TMEM204 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| CLP24/TMEM204 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79652 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 652424 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| C16orf30chromosome 16 open reading frame 30, Claudin-like protein 24, CLP24MGC111564, FLJ20898, transmembrane protein 204 | |
| NS0 mouse myeloma cell line transfected with human CLP24, Accession # Q9BSN7 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction