missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPP Monoclonal Antibody (OTI1G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502061
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.Specifications
| CLPP | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16740 | |
| CLPP | |
| Full length human recombit protein of human CLPP produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G9 | |
| Unconjugated | |
| CLPP | |
| DFNB81, PRLTS3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8192 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction