missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLPP Recombinant Rabbit Monoclonal Antibody (23GB1865), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | CLPP |
|---|---|
| Clone | 23GB1865 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087735
![]() |
Invitrogen™
MA554110 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089388
|
Invitrogen™
MA554111 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLPP Recombinant Monoclonal antibody specifically detects CLPP in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| CLPP | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O88696, Q16740 | |
| 301117, 53895, 8192 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1865 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ATP-dependent Clp protease proteolytic subunit, mitochondrial; ATP-dependent protease ClpAP, proteolytic subunit, human; AU019820; caseinolytic mitochondrial matrix peptidase proteolytic subunit; caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; caseinolytic protease, ATP-dependent, proteolytic subunit homolog; Clpp; ClpP caseinolytic peptidase ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog; D17Wsu160e; DFNB81; endopeptidase Clp; LOW QUALITY PROTEIN: ATP-dependent Clp protease proteolytic subunit, mitochondrial; PRLTS3; putative ATP-dependent Clp protease proteolytic subunit, mitochondrial | |
| A synthesized peptide derived from human CLPP (210-277AA). | |
| Primary | |
| -20°C | |
| CLPP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title