missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPTM1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32114AF405
This item is not returnable.
View return policy
Description
CLPTM1 Polyclonal antibody specifically detects CLPTM1 in Human samples. It is validated for Western Blot
Specifications
| CLPTM1 | |
| Polyclonal | |
| Western Blot | |
| cleft lip and palate associated transmembrane protein 1, cleft lip and palate transmembrane protein 1 | |
| The immunogen this antibody was made to, maps to a region between residue 619 to 669 of human Cleft Lip and Palate Associated Transmembrane Protein 1 using the numbering given in entry NP_001285.1 (GeneID 1209). | |
| 0.1 mL | |
| Cell Biology | |
| 1209 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction